missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAI3 Monoclonal Antibody (409611)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561367
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This p53-target gene encodes a brain-specific angiogenesis inhibitor, a seven-span transmembrane protein, and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities, and may also play a role in angiogenesis.
Specifications
| BAI3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O60242 | |
| Adgrb3 | |
| Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 409611 | |
| Unconjugated | |
| Adgrb3 | |
| A830096D10Rik; ADGRB3; Adhesion G protein-coupled receptor B3; BAI3; Brain-specific angiogenesis inhibitor 3; dJ91B17.1 (brain-specific angiogenesis inhibitor 3); G protein-coupled receptor BAI3; KIAA0550 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction