missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI3 Antibody (409611), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3965M
This item is not returnable.
View return policy
Description
BAI3 Monoclonal antibody specifically detects BAI3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| BAI3 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 409611 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100 | |
| Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242 | |
| 0.1 mL | |
| Angiogenesis, Cancer, GPCR | |
| 577 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction