missing translation for 'onlineSavingsMsg'
Learn More

BAI2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19680-1-AP-150UL

 View more versions of this product

Product Code. 16849154

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to BAI2.

BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
Specifications

Specifications

BAI2
Polyclonal
Unconjugated
ADGRB2
BAI2, brain angiogenesis inhibitor 2
Rabbit
Antigen Affinity Chromatography
RUO
576
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60241
ADGRB2
Peptide
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.