missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550629
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total BAI2 protein.
BAI1, a p53-target gene, encodes brain-specific angiogenesis inhibitor, a seven-span transmembrane protein and is thought to be a member of the secretin receptor family. Brain-specific angiogenesis proteins BAI2 and BAI3 are similar to BAI1 in structure, have similar tissue specificities and may also play a role in angiogenesis.
Specifications
| BAI2 | |
| Polyclonal | |
| Unconjugated | |
| ADGRB2 | |
| Adgrb2; Adhesion G protein-coupled receptor B2; BAI2; Brain-specific angiogenesis inhibitor 2; G protein-coupled receptor brain-specific angiogenesis inhibitor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 230775, 576 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60241, Q8CGM1 | |
| ADGRB2 | |
| Synthetic peptide corresponding to residues near the C terminal of human brain-specific angiogenesis inhibitor 2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction