missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAI1 Antibody (480912) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB49691JF525
This item is not returnable.
View return policy
Description
BAI1 Monoclonal antibody specifically detects BAI1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| BAI1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF | |
| HEK293 human embryonic kidney cell line transfected with human BAI1, Ala31-Thr879, Accession # O14514 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 480912 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience | |
| 575 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction