missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAGE2 Polyclonal Antibody

Description
Immunogen sequence: RLMKEESPVV SWRLEPEDGT ALDVHFVSTL EPLSNAVKRN VPRCIIILVL QEPTPFRISV TSSCFVQNTL TKLLKDRRKM QTVQCATARE TS; Positive Samples: A-431.
BAGE2 (BAGE Family Member 2) is a Protein Coding gene. Candidate gene encoding tumor antigens.
Specifications
Specifications
| Antigen | BAGE2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | BAGE2 |
| Gene Accession No. | Q86Y30 |
| Gene Alias | B melanoma antigen 2; B melanoma antigen family member 2; B melanoma antigen family, member 2; BAGE2; cancer/testis antigen 2.2; cancer/testis antigen family 2, member 2; CT2.2 |
| Gene Symbols | BAGE2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?