Learn More
Invitrogen™ BAGE2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA567594
Product Code. 15836013
  • £440.00 / 100µL
Description
BAGE2 Polyclonal Antibody for Western Blot
BAGE2 (BAGE Family Member 2) is a Protein Coding gene. Candidate gene encoding tumor antigens.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAGE2
Polyclonal
Unconjugated
BAGE2
B melanoma antigen 2; B melanoma antigen family member 2; B melanoma antigen family, member 2; BAGE2; cancer/testis antigen 2.2; cancer/testis antigen family 2, member 2; CT2.2
Rabbit
Antigen affinity chromatography
RUO
85319
-20°C
Liquid
Western Blot
0.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86Y30
BAGE2
Synthesized peptide derived from human BAGE2.