missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAG6 Recombinant Rabbit Monoclonal Antibody (8P4D9)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA542688

Product Code. 17837992

  • £427.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive test controls include: 293T, NIH/3T3, C6, Mouse testis. The target is usually found in the following locations: Cytoplasm, Nucleus, cytosol. Immunogen sequence: KPQPPLSDAY LSGMPAKRRK TMQGEGPQLL LSEAVSRAAK AAGARPLTSP ESLSRDLEAP EVQESYRQQL RSDIQKRLQE DPNYSPQRFP NAQRAFADDP.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAG6
Recombinant Monoclonal
0.49 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P46379, Q6MG49, Q9Z1R2
Bag6
A synthetic peptide corresponding to a sequence within amino acids 1033-1132 of human BAT3/BAG6 (P46379).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
8P4D9
Unconjugated
Bag6
2410045D21Rik; AA408914; BAG 6; BAG family molecular chaperone regulator 6; Bag6; BAG-6; BAT 3; BAT3; BCL2 associated athanogene 6; BCL2-associated athanogene 6; D17H6S52E; D6S52E; DAQB-195H10.3; G3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; protein G3; Protein Scythe; scythe
Rabbit
Affinity Chromatography
RUO
224727, 7917, 94342
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.