missing translation for 'onlineSavingsMsg'
Learn More

BAG3 Antibody (V65P1E8*D2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£159.00 - £302.00

Specifications

Antigen BAG3
Clone V65P1E8*D2
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Applications Western Blot, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18680517
View Documents
Novus Biologicals
NBP2-50112-0.025ml
0.025 mL
£159.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18693068
View Documents
Novus Biologicals
NBP2-50112
0.1 mL
£302.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

BAG3
1 mg/mL
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Endocrinology
PBS
9531
IgG1 κ
Protein A purified
V65P1E8*D2
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Monoclonal
Purified
RUO
Human
BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.