missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG3 Antibody (V65P1E8*D2), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50112AF350
This item is not returnable.
View return policy
Description
BAG3 Monoclonal specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| BAG3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BAG3 | |
| Ovalbumin-conjugated synthetic peptide MTDTPGNPAAP | |
| 0.1 mL | |
| 9531.0 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry | |
| V65P1E8*D2 | |
| Western Blot, ELISA, Immunohistochemistry | |
| BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction