missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG3 Antibody (3D8), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | BAG3 |
|---|---|
| Clone | 3D8 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024605
|
Novus Biologicals
NBP3-26483-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30029802
|
Novus Biologicals
NBP3-26483-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAG3 Monoclonal antibody specifically detects BAG3 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| BAG3 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307 | |
| A synthesized peptide derived from Human BAG3 [UniProt O95817] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3D8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Endocrinology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 9531 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title