missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAG2 Recombinant Rabbit Monoclonal Antibody (23GB7975), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | BAG2 |
|---|---|
| Clone | 23GB7975 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089485
![]() |
Invitrogen™
MA552754 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087518
|
Invitrogen™
MA552755 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAG2 Recombinant Monoclonal antibody specifically detects BAG2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| BAG2 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95816, Q91YN9 | |
| 213539, 690038, 9532 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB7975 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610042A13Rik; BAG cochaperone 2; BAG family molecular chaperone regulator 2; BAG2; BAG-2; BAG-family molecular chaperone regulator-2; BC016230; BCL2 associated athanogene 2; Bcl-2-associated athanogene 2; Bcl2-associated athanogene 2; dJ417I1.2; dJ417I1.2 (BAG-family molecular chaperone regulator 2); KIAA0576; MGC149462; RP3-496N17.2 | |
| A synthesized peptide derived from BAG2 (150-211AA). | |
| Primary | |
| -20°C | |
| BAG2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title