missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19064-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can recognize all the isoforms of BAG1.
The oncogene BCL2 is a membrane protein that blocks a step in a pathway leading to apoptosis or programmed cell death. The protein encoded by this gene binds to BCL2 and is referred to as BCL2-associated athanogene. It enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. At least three protein isoforms are encoded by this mRNA through the use of a non-AUG (CUG) start site, and alternative, downstream, AUG translation initiation sites.Specifications
| BAG1 | |
| Polyclonal | |
| Unconjugated | |
| BAG1 | |
| BAG 1, BAG1, BAG1L speicfic, BAG1M, BAG1S, HAP, RAP46 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12017, 297994, 573 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0K019, Q60739, Q99933 | |
| BAG1 | |
| BAG1 Fusion Protein Ag13555 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction