missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAG1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13441867
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13441867 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13441867 Supplier Invitrogen™ Supplier No. PA527709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: H1299. Predicted reactivity: Mouse (82%), Rat (81%), Pig (88%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The oncogene BCL2 is a membrane protein that blocks a step in a pathway leading to apoptosis or programmed cell death. The protein encoded by this gene binds to BCL2 and is referred to as BCL2-associated athanogene. It enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. At least three protein isoforms are encoded by this mRNA through the use of a non-AUG (CUG) start site, and alternative, downstream, AUG translation initiation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAG1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Bag1
Gene Accession No. Q99933
Gene Alias BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2
Gene Symbols Bag1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Bag1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 573
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.