missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BAG1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149461
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The oncogene BCL2 is a membrane protein that blocks a step in a pathway leading to apoptosis or programmed cell death. The protein encoded by this gene binds to BCL2 and is referred to as BCL2-associated athanogene. It enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. At least three protein isoforms are encoded by this mRNA through the use of a non-AUG (CUG) start site, and alternative, downstream, AUG translation initiation sites.
Specifications
| BAG1 | |
| Polyclonal | |
| Unconjugated | |
| Bag1 | |
| BAG family molecular chaperone regulator 1; Bag1; BAG-1; BCL2 associated athanogene 1; Bcl-2 associating athanogene-1 protein; BCL2-associated athanogene; bcl-2-associated athanogene 1; BCL2-associated athanogene 1; Bcl-2-binding protein; glucocortoid receptor-associated protein RAP46; HAP; Rap46; receptor-associated protein, 46-KD; RP11-326F20.2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 12017 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q60739 | |
| Bag1 | |
| E. coli-derived recombinant mouse Bag-1. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction