missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bag-1 Antibody (RM310), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77409
This item is not returnable.
View return policy
Description
Bag-1 Monoclonal antibody specifically detects Bag-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Bag-1 | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| RM310 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000 | |
| BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 | |
| A peptide corresponding to residues near the N-terminus of human Bag-1 | |
| 100 μg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 573 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion