missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bag-1 Polyclonal specifically detects Bag-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Bag-1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | BAG family molecular chaperone regulator 1, BAG-1, Bcl-2 associating athanogene-1 protein, BCL2-associated athanogene, Bcl-2-associated athanogene 1, Bcl-2-binding protein, glucocortoid receptor-associated protein RAP46,46-KD, HAP, RAP46 |
| Gene Symbols | BAG1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 26-45 (NERYDLLVTPQQGNSEPVVQD) of mouse BAG-1 was used as immunogen; GenBank no. AAH03722.1. This sequence is 100% conserved in mouse BAG-1L; GenBank no. NP_033866.3. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?