missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bafilomycin A1, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases
- It is a macrolide antibiotic, and shows fungicide activity
Application
- Bafilomycin A1 has antibacterial, antifungal, and antineoplastic activity
- It can be immunosuppressive
- It can prevent maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes
- It is a potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase)
Specifications
Specifications
| Type | Bafilomycin A1 |
| Physical Form | Powder/Solid |
| CAS | 88899-55-2 |
| Color | White |
| Quantity | 0.1 mg |
| Assay Percent Range | 95% |
| Molecular Formula | C35H58O9 |
| MDL Number | MFCD06795130 |
| Solubility Information | Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ethanol, dmso |
| InChI Key | XDHNQDDQEHDUTM-XRCNURIOSA-N |
| Show More |
Safety and Handling
Product Identifier
- Bafilomycin A1
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction