missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Bafilomycin A1
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Bafilomycin A1 is a highly potent, selective inhibitor of vacuolar H+-ATPases (IC50 = 0.6 - 1.5 nM in bovine chromaffin granules). Selective for v-ATPase over other ATP hydrolyzing enzymes such as F-ATPases and the H+/K+-ATPase (P-ATPase). Thought to inhibit autophagy either by blocking autophagosome-lysosome fusion (in H4IIE cells), or by blocking lysosomal degradation. Bafilomycin A1 accelerates wound healing in db/db mice. Low dose Bafilomycin A1 attenuates patient-derived CD34+CD19+ leukemia stem cells (LSC) and inhibits LSC proliferation in an animal model. Bafilomycin A1 impairs Zika virus infection in vitro.
Specifications
Specifications
| Chemical Name or Material | (3Z,5E,7R,8S,9S,11E,13E,15S,16R)-8-Hydroxy-16-[(1S,2R,3S)-2-hydroxy-1-methyl-3-[(2R,4R,5S,6R)-tetrahydro-2,4-dihydroxy-5-methyl-6-(1-methylethyl)-2H-pyran-2-yl]butyl]-3,15-dimethoxy-5,7,9,11-tetramethyloxacyclohexadeca-3,5,11,13-tetraen-2-one |
| CAS | 88899-55-2 |
| Quantity | 100 μg |
| Target | H+-ATPase Inhibitors |
| Molecular Formula | C35H58O9 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction