missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Mouse anti-Human, Clone: BAFFR/1558, Novus Biologicals™
Mouse Monoclonal Antibody
£215.00 - £396.00
Specifications
| Antigen | BAFFR/TNFRSF13C |
|---|---|
| Clone | BAFFR/1558 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326094
|
Novus Biologicals
NBP3-07544-20UG |
20 μg |
£215.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18397412
|
Novus Biologicals
NBP3-07544-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| BAFFR/TNFRSF13C | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant full-length human BAFFR/TNFRSF13C protein | |
| Primary | |
| Store at 4°C. |
| BAFFR/1558 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 115650 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title