missing translation for 'onlineSavingsMsg'
Learn More

BAFFR/TNFRSF13C Mouse anti-Human, Clone: BAFFR/1558, Novus Biologicals™

Product Code. 18397412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397412 100 μg 100µg
18326094 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18397412 Supplier Novus Biologicals Supplier No. NBP307544100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BAFFR/1558
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Recombinant full-length human BAFFR/TNFRSF13C protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.