missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Mouse anti-Human, Clone: BAFFR/1557, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07543-100ug
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| BAFFR/TNFRSF13C | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| BAFFR/1557 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant full-length human BAFFR/TNFRSF13C protein | |
| 100 μg | |
| Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
| 115650 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto