missing translation for 'onlineSavingsMsg'
Learn More

BAFFR/TNFRSF13C Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18418002 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418002 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418002 Supplier Novus Biologicals Supplier No. NBP177234AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BAFFR/TNFRSF13C Polyclonal antibody specifically detects BAFFR/TNFRSF13C in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAFFR/TNFRSF13C
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of human BAFF Receptor The peptide sequence is identical between human and mouse origin. The immunogen is located within the last 50 amino acids of BAFF Receptor. Amino Acid Squence: GSTELVTTKTAGPEQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.