missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BAFFR/TNFRSF13C Antibody (5A9B10), Novus Biologicals™
Mouse Monoclonal Antibody
£217.00 - £389.00
Specifications
| Antigen | BAFFR/TNFRSF13C |
|---|---|
| Clone | 5A9B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30037537
|
Novus Biologicals™
NBP3-27126-0.1ml |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028375
|
Novus Biologicals™
NBP3-27126-0.025ml |
0.025 mL |
£217.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| BAFFR/TNFRSF13C | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
| PBS | |
| 115650 | |
| IgG2b | |
| Protein G purified |
| 5A9B10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C | |
| Purified recombinant fragment of human BAFFR/TNFRSF13C (AA: extra 1-78) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title