missing translation for 'onlineSavingsMsg'
Learn More

PeproTech BAFF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Brand:  PeproTech 500-P163GBT-50UG

 View more versions of this product

Product Code. 17824063

  • £228.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: AVQGPEETVT QDCLQLIADS ETPTIQKGSY TFVPWLLSFK RGSALEEKEN KILVKETGYF FIYGQVLYTD KTYAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PRENAQISLD GDVTFFGALK LL. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human BAFF. Anti-Human BAFF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBAFF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BAFF (500-P163G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBAFF. Western Blot: To detect hBAFF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBAFF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P163GBT-1MG will be provided as 2 x 500 μg

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAFF
Polyclonal
Biotin
TNFSF13B
ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4
Goat
Antigen affinity chromatography
RUO
10673
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q9Y275
TNFSF13B
E.coli-derived Recombinant Human BAFF.
50 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.