missing translation for 'onlineSavingsMsg'
Learn More

BAFF/BLyS/TNFSF13B Antibody (H4-C7), PE/Cy5.5, Novus Biologicals™

Product Code. 18252383 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18252383 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18252383 Supplier Novus Biologicals Supplier No. NBP104265PECY55

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAFF/BLyS/TNFSF13B
Applications Flow Cytometry
Classification Monoclonal
Clone H4-C7
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Gene Symbols TNFSF13B
Host Species Mouse
Immunogen Recombinant human BLyS (134-285aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10673.0
Test Specificity Anti-human BLyS mAb, clone H4-C7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BLyS protein.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG3 κ
Show More Show Less
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.