Learn More
Invitrogen™ BAF57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592420
Description
Immunogen sequence: MSKRPSYAPP PTPAPATQMP STPGFVGYNP YSHLAYNNYR LGGNPGTNSR VTASSGITIP KPPKPPDKPL MPYMRYSRKV WDQVKASNPD LKLWEIGKII GGMWRDLTDE EKQEYLNEYE AEKIEYNESM KAYHNSPAYL AYINAKSRAE AALEEESRQR QSRMEKGEPY MSIQPAEDPD DYDDGFSMKH TATARFQRNH RLISEILSES VVPDVRSVVT TARMQVLKRQ VQSLMVHQRK LEAELLQIEE RHQEKKRKFL ESTDSFNNEL KRLCGLKVEV DMEKIAAEIA QAEEQARKRQ EEREKEAAEQ AERSQSSIVP EEEQAANKGE EKKDDENIPM ETEETHLEET TESQQNGEEG TSTPEDKESG QEGVDSMAEE GTSDSNTGSE SNSATVEEPP TDPIPEDEKK E; Positive Samples: U-251MG, Jurkat; Cellular Location: Nucleus.
The protein encoded by this gene is part of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. The encoded protein, either alone or when in the SWI/SNF complex, can bind to 4-way junction DNA, which is thought to mimic the topology of DNA as it enters or exits the nucleosome. The protein contains a DNA-binding HMG domain, but disruption of this domain does not abolish the DNA-binding or nucleosome-displacement activities of the SWI/SNF complex. Unlike most of the SWI/SNF complex proteins, this protein has no yeast counterpart.
Specifications
| BAF57 | |
| Polyclonal | |
| Unconjugated | |
| Smarce1 | |
| 2810417B20Rik; 5830412H02Rik; 9030408N19Rik; Baf27; BAF57; BRG1-associated factor 57; chromatin remodeling complex BRG1-associated factor 57; CSS5; RGD1304726; SMARCE 1; Smarce1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1; SWI/SNF-related matrix-associated actin-dependent regulator chromatin subfamily E member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57376, 6605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54941, Q969G3 | |
| Smarce1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-411 of human BAF57/BAF57/SMARCE1 (NP_0030703). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.