missing translation for 'onlineSavingsMsg'
Learn More

BAF53A Rabbit anti-Human, Mouse, Rat, Clone: 5U6J8, Novus Biologicals™

Product Code. 18329195
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329195 20 μg 20µL
18380533 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18329195 Supplier Novus Biologicals Supplier No. NBP31665920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BAF53A Monoclonal antibody specifically detects BAF53A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF53A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5U6J8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias actin-like 6A, actin-like protein 6A, actin-related protein 4,53 kDa BRG1-associated factor A, Actin-related protein Baf53a, Actl6, Arp4, ArpNbeta, BAF complex 53 kDa subunit, Baf53a, BAF53AACTL6, BAF53MGC5382, BRG1-associated factor 53A, hArpN beta, INO80 complex subunit K, INO80KARPN-BETA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 201-290 of human BAF53A (O96019).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 86
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.