Learn More
Invitrogen™ BAF53A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519421
Description
PA5-19421 targets BAF53A in IF, IHC (F), and WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-19421 immunogen is synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Human BAF53A. PA5-19421 detects BAF53A which has a predicted molecular weight of approximately 53 kDa.
BAF53A is a family member of actin-related proteins (ARPs), which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a 53 kDa subunit protein of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. Together with beta-actin, it is required for maximal ATPase activity of BRG1, and for the association of the BAF complex with chromatin/matrix. Three transcript variants that encode two different protein isoforms have been described.
Specifications
| BAF53A | |
| Polyclonal | |
| Unconjugated | |
| Actl6a | |
| 2810432C06Rik; 53 kDa BRG1-associated factor A; actin like 6A; actin-like 6; actin-like 6A; actin-like 6B; actin-like protein 6A; actin-related protein 4; actin-related protein Baf53a; Actl6; Actl6a; ACTL6B; AI851094; ARP4; ArpNbeta; ARPN-BETA; BAF complex 53 kDa subunit; BAF53; Baf53a; BAF53B; BRG1/brm-associated factor 53A; BRG1-associated factor 53A; C79802; hArpN alpha; hArpN beta; INO80 complex subunit K; INO80K; MGC5382; MGC5382 Arp4; RGD1307747 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361925, 56456, 86 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| O96019, Q9Z2N8 | |
| Actl6a | |
| Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Human BAF53A. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.