Learn More
Specac™ BaF2 Sealed Heatable Liquid Static Cell
Product Code. 15220208
Encompass_Preferred
Change view
Click to view available options
Type:
BaF2 Sealed Heatable Liquid Static Cell
Product Code. Type
15220208 BaF2 Sealed Heatable Liquid Static Cell
1 options
Product Code. 15220208 Supplier Specac™ Supplier No. GS20503
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This heatable liquid cell is designed for high sensitivity ATR measurements with FTIR or dispersive instruments. This is a sealed cell with BaF2 windows with a choice of different pathlengths available ranging from 0.025mm up to 1.0mm.
Specifications
Type BaF2 Sealed Heatable Liquid Static Cell
Description Pathlengths available ranging from 0.025 mm up to 1.0 mm