Learn More
Specac™ BaF2 Sealed Heatable Liquid Flow Cell VTCH Compatible
Product Code. 15280218
Encompass_Preferred
Change view
Click to view available options
Type:
BaF2 Sealed Heatable Liquid Flow Cell
Product Code. Type
15280218 BaF2 Sealed Heatable Liquid Flow Cell
1 options
Product Code. 15280218 Supplier Specac™ Supplier No. GS20563
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
This cell is designed for use in the Specac variable temperature cell GS21525. This is a sealed heatable liquid flow cell with BaF2 windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type BaF2 Sealed Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm