Learn More
Specac™ BaF2 Demountable Heatable Liquid Flow Cell VTCH Compatible
Product Code. 15230238
Encompass_Preferred
Click to view available options
Type:
BaF2 Demountable Heatable Liquid Flow Cell
Product Code. 15230238
Brand: Specac™ GS20593
This cell is designed for use in the Specac variable temperature cell GS21525. This is a demountable heatable liquid flow cell with BaF2 windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type BaF2 Demountable Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm