missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAF180/PB1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79832CL1
This item is not returnable.
View return policy
Description
BAF180/PB1 Polyclonal antibody specifically detects BAF180/PB1 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Specifications
| BAF180/PB1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| BAF180PB1BRG1-associated factor 180, hPB1, MGC156155, MGC156156, polybromo 1, Polybromo-1D, protein polybromo-1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human polybromo 1 (BRG1-associated factor 180) using the numbering given in entry AAP34197.1 (GeneID 55193). | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 55193 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction