missing translation for 'onlineSavingsMsg'
Learn More

BAF180/PB1 Antibody (CL0331), Novus Biologicals™

Mouse Monoclonal Antibody

£292.00 - £435.00

Specifications

Antigen BAF180/PB1
Clone CL0331
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18426491
View Documents
Novus Biologicals
NBP2-30673-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18145054
View Documents
Novus Biologicals
NBP2-30673
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BAF180/PB1 Monoclonal specifically detects BAF180/PB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

BAF180/PB1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
Mouse
Human
BAF180PB1BRG1-associated factor 180, hPB1, MGC156155, MGC156156, polybromo 1, Polybromo-1D, protein polybromo-1
PBRM1
IgG1
Protein A purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
CL0331
Monoclonal
Purified
RUO
Q86U86
55193
This antibody was developed against a recombinant protein corresponding to amino acids: KMEEYDDVNLLTADFQLLFNNAKSYYKPDSPEYKAACKLWDLYLRTRNEFVQKGEADDEDDDEDGQDNQGTVTEGSSPAYLKEILEQLLEAIVVATNPSGRLISELFQKLPSKVQYPDYYAIIKEPIDLKTIAQRIQNGSYKSIHA
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.