missing translation for 'onlineSavingsMsg'
Learn More

Bad, p Ser112 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£193.00 - £320.00

Specifications

Antigen Bad (p Ser112)
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18224651
View Documents
Novus Biologicals
NB100-81807-0.025ML
0.025 mL
£193.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18759033
View Documents
Novus Biologicals
NB100-81807
0.05 mL
£320.00
0.05mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bad Polyclonal specifically detects Bad in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Bad (p Ser112)
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Polyclonal
Rabbit
Phospho Specific
Q92934
572
IgG
Affinity Purified
23 kDa
1.0 mg/mL
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primate, Mouse, Human
BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter
BAD
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.