Learn More
Baculovirus Envelope gp64 protein Antibody (AcV1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00208PCP
This item is not returnable.
View return policy
Description
Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.
Specifications
| Flow Cytometry | |
| Affinity Purified |