missing translation for 'onlineSavingsMsg'
Learn More

Baculovirus Envelope gp64 protein Antibody (AcV1), PE/Atto594, Novus Biologicals™

Product Code. 18405111 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405111 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405111 Supplier Novus Biologicals Supplier No. NBP200208PEATT594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Baculovirus Envelope gp64 protein Monoclonal antibody specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Baculovirus Envelope gp64 protein
Applications Flow Cytometry
Classification Monoclonal
Clone AcV1
Conjugate PE/ATTO 594
Formulation PBS
Gene Alias ACNVgp129, AcOrf-128
Host Species Mouse
Immunogen The immunogen for this antibody is baculovirus envelope gp64 protein.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.