missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Baculovirus Envelope gp64 protein Antibody (AcV1), DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Baculovirus Envelope gp64 protein Monoclonal antibody specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Baculovirus Envelope gp64 protein |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AcV1 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ACNVgp129, AcOrf-128 |
| Host Species | Mouse |
| Immunogen | The immunogen for this antibody is baculovirus envelope gp64 protein. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?