Learn More
Gibco™ Bacto™ Proteose Peptone No. 3
Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media.
Brand:  Gibco™ 211692
Additional Details : Weight : 51.30000kg
 View more versions of this product
Product Code. 16239751
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Specifications
LUO, MUO
Mammalian, Insect, Microbial
Dehydrated Culture Media
Bacto™
Animal Origin
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Microbiological and mammalian cell culture media
50 kg