missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Malt Extract

The water-soluble portion of malted barley.

Brand:  Gibco™ 218630

Additional Details : Weight : 0.54210kg

 View more versions of this product

Product Code. 16289841

  • £76.25 / 500g
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Bacto Malt Extract is the water-soluble portion of malted barley
  • Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium
  • It has been used successfully in microbial fermentation applications
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications

Specifications

LUO
Powder
Bottle
Bacto™
Animal origin-free, malted barley
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Malt Extract
Microbial fermentation applications
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers