missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (05), FITC, Invitrogen™

Mouse Recombinant Monoclonal Antibody
£151.00 - £405.00
Specifications
| Antigen | Bacteriophage M13 |
|---|---|
| Clone | 05 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019222
![]() |
Invitrogen™
MA552322 |
100 Tests |
£405.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019626
|
Invitrogen™
MA552323 |
25 Tests |
£151.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bacteriophage M13 Recombinant Monoclonal antibody specifically detects Bacteriophage M13 in Virus samples. It is validated for Flow CytometrySpecifications
| Bacteriophage M13 | |
| 0.1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| M13 Bacteriophage. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 05 | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| Virus | |
| Enterobacteria phage M13 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title