missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bacteriophage M13 Monoclonal Antibody (K70008_8E2), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | Bacteriophage M13 |
|---|---|
| Clone | K70008_8E2 |
| Concentration | 0.77 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30195383
|
Invitrogen™
MA555990 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211028
|
Invitrogen™
MA556370 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bacteriophage M13 Monoclonal antibody specifically detects Bacteriophage M13 in Chemical samples. It is validated for Western BlotSpecifications
| Bacteriophage M13 | |
| 0.77 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| M13 bacteriophage. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| K70008_8E2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Enterobacteria phage M13 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title