Learn More
Thermo Scientific™ Bacteriological Peptone No. 2
Description
Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralised form are very nutritious all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. They are specially prepared to be compatible with other refined culture media or fermentation ingredients.
The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles.
The neutralised form evolved from the original to meet those occasions when a slightly higher pH is required. Both when reconstituted give a solution free of haze, cloudiness or precipitation.
LP0137 is manufactured using beef from New Zealand.
Some key benefits of using dehydrated culture media include:
Ease of preparation: Simply add water to rehydrate the media.
Consistency: Ensures uniformity in microbial growth conditions.
Storage: Can be stored at room temperature for extended periods.
Specifications
Specifications
| Content And Storage | Dehydrated Powder, Ambient & Dry Storage |
| Form | Dehydrated Powder |
| Manufacturing Origin | United Kingdom |
| Media Type | Bacteriological Peptone No. 2 |
| Product Type | Media Additive |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbiological Tests |
| For Use With (Application) | Culture media or fermentation processes |
| Quantity | 500 g |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.