missing translation for 'onlineSavingsMsg'
Learn More

Bactericidal Disinfectant Wipes

Product Code. 12869297
Encompass_Preferred
Click to view available options
Dimensions (L x W):
200 x 220 mm
Unit Size:
0.2kg
This item is not returnable. View return policy

Product Code. 12869297

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ideal for use in laboratories or clinical environments where rapid disinfection of clinical surfaces is essential. Bactericidal Disinfectant Wipes are supplied in an easy dispense tub design.

  • Manufactured from white, non woven material which is perforated for ease of dispensing
  • 70% v/v denatured ethanol B and chlorhexidine digluconate, 1% w/v solution
  • Fast, effective decontamination of all surfaces
  • Superior to isopropyl alcohol
  • Easy dispense tub design

Specifications

Product Type Bactericidal Disinfectant Wipes
Sterility Non-sterile
Appearance White
Composition 70% V/V denatured ethanol B and chlorhexidine digluconate, 1.25% W/V solution, superior to isopropyl
Container Type Plastic tub dispenser
Dimensions (L x W) 200 x 220 mm
For Use With (Application) Ideal for use in laboratories or clinical environments where rapid disinfection of clinical surfaces is essential
Quantity 160 Wipes
compliance-icons
Product Identifier
  • ShermondWipe 7200
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : ETHANOL, DC ACID
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.