Learn More
Cytiva (formerly Pall Lab) Bacterial Air Vents
Product Code. 17184271
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 24
Product Code. 17184271
Brand: Cytiva (formerly Pall Lab) 4210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use as vents for receiving vessels or as sterile breathing barriers.
  • Economical and convenient depth filter for venting applications
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Glass Laminate
Pore Size 1 μm
Sterility Sterile
Filtration Area 7.5 cm2
Inlet Style Stepped Hose Barb
Outlet Style Stepped Hose Barb
Quantity 24/Pk.
Diameter (Metric) 37 mm