missing translation for 'onlineSavingsMsg'
Learn More

Cytiva bacteria genomicPrep Mini Spin Kit

Product Code. 10546485 Shop All Cytiva Products
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Unit Size:
250 reactions
Each
This item is not returnable. View return policy

Product Code. 10546485

Brand: Cytiva 28904259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

illustra bacteria genomicPrep Mini spin columns prepacked with novel silica membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; a full booklet with a detachable, quick reference protocol card

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria

The illustra Bacteria genomicPrep Mini Spin Kit can be used with a wide range of bacterial strains such as DH5a, TOP10, JM109, Bacillus subtilis etc. The kit produces highly pure and intact genomic DNA with OD260/280 values of 1.75-1.85. Use different elution volumes to isolate genomic DNA at concentrations suitable for most downstream applications including restriction enzyme digestion, RT-PCR, and long PCR.

The method uses a lysis solution in combination with proteinase K to gently release genomic DNA into solution from bacteria cells. The DNA is deproteinated in the extraction solution and the genomic DNA is then bound onto a silica column in the presence of chaotropic solution. Contaminants are removed during wash steps and the purifed DNA is eluted with prewarmed TE solution.

  • Total DNA extraction time under one hour with reduced lysis time
  • Can be used to purify 4 to 8μg of genomic DNA from 2 × 109 bacteria cells with input amounts ranging from 1 × 109 to 4 × 109 bacteria cells
  • Spin columns prepacked with a silica membrane—provide excellent reproducibility and consistency
  • Validated in several downstream applications including RT PCR, long PCR, and restriction digest
  • Color-coded caps for ease of use

TRUSTED_SUSTAINABILITY

Specifications

Description illustra bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.
For Use With (Application) For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteriaPurified gDNA is suitable for molecular biology applications including cloning, restriction enzyme digestion, PCR and genotyping
Includes 30mg Proteinase K, 3mL Lysis Buffer type 2, 0.6mL Lysis Buffer type 3, 60mL Lysis Buffer type 4, 6mL Wash Buffer type 6, 12mL Elution Buffer type 5, 50 Illustra™ bacteria mini columns, 50 collection tubes
No. of Reactions 250 Preps
Product Type illustra bacteria genomicPrep Mini Spin Kit
Purity 1.7
Sample Volume 2 x 109 cells [A600 = 2.0]
Test Time 40 min [Gram-negaitive Bacteria], 55 min [Gram-positive Bacteria]
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : proteases
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.