missing translation for 'onlineSavingsMsg'
Learn More

Thomil™ Bacter Quat pH Neutral Bactericidal, Fungicidal and Virucidal Cleaner

Product Code. 17004297
Encompass
Click to view available options
Volume (Metric):
750 mL
4 L
Packaging:
Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish
Unit Size:
Pack of 12
Pack of 4
This item is not returnable. View return policy
Contains: Benzalkonium Chloride (4.56%), For Professional Use Only

Product Code. 17004297

Brand: Thomil™ LSLG064EX

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Contains: Benzalkonium Chloride (4.56%), For Professional Use Only

Bacter Quat neutral pH cleaner provides triple action power by cleaning, disinfecting, and deodorizing. The solution can be applied to any surface without the risk of damage that is typically caused by cleaners.

  • Triple action power cleans, disinfects, and deodorizes at once
  • For the disinfection of all types of general surfaces, especially in the food industry
  • Tested efficiency against coronavirus
  • UNE-EN 14476 against Norovirus, Adenovirus and Rotavius in 15% solution after 15 minutes of contact
  • UNE-EN 13697 bactericidal, fungicidal efficiency in 5% solution after 5 minutes of contact
  • Removes dirt without damaging materials, therefore it can be used as the only product for cleaning and disinfection in places of business
  • Registered in DGSP of Spain, NHS of UK, ANSES of France, DGS and DGAV of Portugal, and PCS of Ireland

Specifications

Product Type Chemical Disinfectant
Form Liquid
pH 7 to 8
Sterility Non-sterile
Appearance Blue
Boiling Point 100°C
Certifications/Compliance UNE-EN 13697, UNE-EN 14476
Composition Quaternary Ammonium Compounds, Benzyl-C12-16-alkyldimethyl (4.56%), Chlorides
Concentration 1 - <5%
Container Type Canister
Content And Storage 5°C to 40°C
Description Bacter Quat pH Neutral Bactericida, Fungicide and Virucide Cleaner
Dilution 5% to 15%
Disposable Reusable
Filtered Unfiltered
For Use With (Application) Cleaning, Disinfecting, and Deodorizing All Types of Surfaces
Packaging Labeling: English, French, German, Italian, Portuguese, Russian, Arabic, and Spanish
Quantity 4 Units/Box
Volume (Metric) 4 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • X4 4L BACTER QUAT PH Neutral B
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : aqueous mixture of quarternary ammonia compound
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.