missing translation for 'onlineSavingsMsg'
Learn More

BD Bactec™ Media: Myco/F Lytic

Product Code. p-4907892
Click to view available options
Quantity:
50/Pk.
This item is not returnable. View return policy

Product Code. 17352603

Brand: BD 442288

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with BACTEC 9240, 9120, and 9050 Systems

  • Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
  • Fluorescent series bar-coded blood culture media
  • Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
  • Vials ready to use as received—require no reconstitution or dilution
  • Detection is limited to microorganisms that will grow in a particular type of medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 25°C
Form Liquid
Media Type Myco/F Lytic
Product Type Microbiology Media
Formulation Processed Water 40mL, 7H9 middlebrook broth base without phosphate Salts 0.12% w/v, Brain heart infusion 0.5% w/v, Casein Hydrolysate 0.10% w/v, Supplement H 0.10% w/v, Inositol 0.05% w/v, Glycerol 0.10% w/v, Sodium Polyanetholsulfonate 0.025% w/v, Polyso
For Use With (Equipment) Bactec™ 9240, 9120 and 9050 System
Packaging Type Vial
Quantity 50/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.