missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Used in a qualitative procedure for the culture and recovery of anaerobic microorganisms from blood
- Fluorescent series bar-coded blood culture media
- Each vial contains a chemical sensor which can detect increases in CO2 produced by the growth of microorganisms
- Vials ready to use as received—require no reconstitution or dilution
- Detection is limited to microorganisms that will grow in a particular type of medium
Specifications
Specifications
| Content And Storage | 2 to 25°C |
| Form | Liquid |
| Media Type | Myco/F Lytic |
| Product Type | Blood Culture Medium |
| Formulation | Processed Water 40mL, 7H9 middlebrook broth base without phosphate Salts 0.12% w/v, Brain heart infusion 0.5% w/v, Casein Hydrolysate 0.10% w/v, Supplement H 0.10% w/v, Inositol 0.05% w/v, Glycerol 0.10% w/v, Sodium Polyanetholsulfonate 0.025% w/v, Polyso |
| For Use With (Equipment) | Bactec™ 9240, 9120 and 9050 System |
| Packaging Type | Vial |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?