missing translation for 'onlineSavingsMsg'
Learn More

Baclofen, TRC

Product Code. 30334225
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334225 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334225 Supplier Toronto Research Chemicals Supplier No. TRCB1080001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Baclofen
CAS 1134-47-0
Molecular Formula C10 H12 Cl N O2
Quantity 1 g
Synonym Baclofen, (3RS)-4-Amino-3-(4-chlorophenyl)butanoic acid, Benzenepropanoic acid, β-(aminomethyl)-4-chloro- (9CI, ACI), Hydrocinnamic acid, β-(aminomethyl)-p-chloro- (7CI, 8CI), β-(Aminomethyl)-4-chlorobenzenepropanoic acid (ACI), (±)-4-Amino-3-(4-chlorophenyl)butanoic acid, (±)-Baclofen, 4-Amino-3-(4-chlorophenyl)butanoic acid, 4-Amino-3-(4-chlorophenyl)butyric acid, 4-Amino-3-(p-chlorophenyl)butyric acid, DL-4-Amino-3-p-chlorophenylbutanoic acid, DL-Baclofen, Ba 34647, Baclon, C 34647Ba, CIBA Ba 34647, dl-Baclofen, Lioresal, Lyvispah, MeSH ID: D001418, β-(4-Chlorophenyl)-γ-aminobutyric acid, β-(Aminomethyl)-p-chlorohydrocinnamic acid, β-(p-Chlorophenyl)-γ-aminobutyric acid, β-p-Chlorophenyl-GABA
InChI Formula InChI=1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)
SMILES NCC(CC(=O)O)c1ccc(Cl)cc1
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
Molecular Weight (g/mol) 213.66
Formula Weight 213.0557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.